missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MIOX Protein

A cDNA sequence encoding the MIOX was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12699-1mg


Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15931269

  • 7540.00 € / 1mg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

MIOX Protein
Research Use Only
Recombinant Protein
E. coli
MKHHHHHHASMKVTVGPDPS LVYRPDVDPE VAKDKASFRN YTSGPLLDRV FTTYKLMHTH QTVDFVRSKH AQFGGFSYKK MTVMEAVDLL DGLVDESDPD VDFPNSFHAF QTAEGIRKAH PDKDWFHLVG LLHDLGKVLA LFGEPQWAVV GDTFPVGCRP QASVVFCDST FQDNPDLQDP RYSTELGMYQ PHCGLDRVLM SWGHDEYMYQ VMKFNKFSLP PEAFYMIRFH SFYPWHTGRD YQQLCSQQDL AMLPWVREFN KFDLYTKCPD LPDVDKLRPY YQGLIDKYCP GILSW
Greater than 95.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
MIOX was filtered (0.4 microm) and lyophilized in 20mM Tris buffer, 50mM NaCl and 5% (w/v) trehalose, pH 7.5.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers