missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Mitochondrial import receptor subunit TOM34 Protein

A DNA sequence encoding the Homo sapiens (Human) Mitochondrial import receptor subunit TOM34, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7602-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16049680

  • 2285.00 € / 1mg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_006800.2
Mitochondrial import receptor subunit TOM34 Protein
Research Use Only
TOMM34
Recombinant Protein
E. coli
MAPKFPDSVE ELRAAGNESF RNGQYAEASA LYGRALRVLQ AQGSSDPEEE SVLYSNRAAC HLKDGNCRDC IKDCTSALAL VPFSIKPLLR RASAYEALEK YPMAYVDYKT VLQIDDNVTS AVEGINRMTR ALMDSLGPEW RLKLPSIPLV PVSAQKRWNS LPSENHKEMA KSKSKETTAT KNRVPSAGDV EKARVLKEEG NELVKKGNHK KAIEKYSESL LCSNLESATY SNRALCYLVL KQYTEAVKDC TEALKLDGKN VKAFYRRAQA HKALKDYKSS FADISNLLQI EPRNGPAQKL RQEVKQNLH
Greater than 80% as determined by SDS-PAGE.
10953
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers