missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MMAB Protein

A cDNA sequence encoding the MMAB was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12702-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15931279

  • 120.00 € / 5µg
Estimated Shipment: 20-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

MMAB Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
MMAB 1 mg/ml protein solution contains 20mM Tris pH 7.5 and 10% glycerol.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MQSRGPQGVE DGDRPQPSSK TPRIPKIYTK TGDKGFSSTF TGERRPKDDQ VFEAVGTTDE LSSAIGFALE LVTEKGHTFA EELQKIQCTL QDVGSALATP CSSAREAHLK YTTFKAGPIL ELEQWIDKYT SQLPPLTAFI LPSGGKISSA LHFCRAVCRR AERRVVPLVQ MGETDANVAK FLNRLSDYLF TLARYAAMKE GNQEKIYKKN DPSAESEGL
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers