missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MNDA Protein
A cDNA sequence encoding the MNDA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12711-1mg
Specifications
MNDA Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MVNEYKKILL LKGFELMDDY HFTSIKSLLA YDLGLTTKMQ EEYNRIKITD LMEKKFQGVA CLDKLIELAK DMPSLKNLVN NLRKEKSKVA KKIKTQEKAP VKKINQEEVG LAAPAPTARN KLTSEARGRI PVAQKRKTPN KEKTEAKRNK VSQEQSKPPG PSGASTSAAV DHPPLPQTSS STPSNTSFTP NQETQAQRQV DARRNVPQND PVTVVVLKAT APFKYESPEN GKSTMFHATV ASKTQYFHVK VFDINLKEKF VRKKVITISD YSECKGVMEI KEASSVSDFN QNFEVPNRII EIANKTPKIS QLYKQASGTM VYGLFMLQKK SVHKKNTIYE IQDNTGSMDV VGSGKWHNIK CEKGDKLRLF CLQLRTVDRK LKLVCGSHSF IKVIKAKKNK EGPMNVN | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The MNDA solution contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT and 10% glycerol. |