missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MOB1B Protein
A cDNA sequence encoding the MOB1B was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12712-5ug
Additional Details : Weight : 0.01000kg
Specifications
MOB1B Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSFLFGS RSSKTFKPKK NIPEGSHQYE LLKHAEATLG SGNLRMAVML PEGEDLNEWV AVNTVDFFNQ INMLYGTITD FCTEESCPVM SAGPKYEYHW ADGTNIKKPI KCSAPKYIDY LMTWVQDQLD DETLFPSKIG VPFPKNFMSV AKTILKRLFR VYAHIYHQHF DPVIQLQEEA HLNTSFKHFI FFVQEFNLID RRELAPLQEL IEKLTSKDR | |
Greater than 95% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MOB1B protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 30% glycerol and 1mM DTT. |