missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human mRNA export factor Protein

Product Code. p-7153369
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16014310 10 μg 10µg
16054310 50 μg 50µg
16004310 100 μg 100µg
16034310 200 μg 200µg
16044310 500 μg 500µg
16024310 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16014310 Supplier enQuireBio™ Supplier No. QP6580ec10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) mRNA export factor, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001015885.1
Gene ID (Entrez) 8480
Name mRNA export factor Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol RAE1
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag GST
Sequence MSLFGTTSGF GTSGTSMFGS ATTDNHNPMK DIEVTSSPDD SIGCLSFSPP TLPGNFLIAG SWANDVRCWE VQDSGQTIPK AQQMHTGPVL DVCWSDDGSK VFTASCDKTA KMWDLSSNQA IQIAQHDAPV KTIHWIKAPN YSCVMTGSWD KTLKFWDTRS SNPMMVLQLP ERCYCADVIY PMAVVATAER GLIVYQLENQ PSEFRRIESP LKHQHRCVAI FKDKQNKPTG FALGSIEGRV AIHYINPPNP AKDNFTFKCH RSNGTNTSAP QDIYAVNGIA FHPVHGTLAT VGSDGRFSFW DKDARTKLKT SEQLDQPISA CCFNHNGNIF AYASSYDWSK GHEFYNPQKK NYIFLRNAAE ELKPRNKK
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 80% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.