missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MRPS2 Protein
A cDNA sequence encoding the MRPS2 was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Name | MRPS2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15931369
|
enQuireBio™
QP12732-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15921369
|
enQuireBio™
QP12732-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15911369
|
enQuireBio™
QP12732-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
MRPS2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The MRPS2 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMATSSAA LPRILGAGAR APSRWLGFLG KATPRPARPS RRTLGSATAL MIRESEDSTD FNDKILNEPL KHSDFFNVKE LFSVRSLFDA RVHLGHKAGC RHRFMEPYIF GSRLDHDIID LEQTATHLQL ALNFTAHMAY RKGIILFISR NRQFSYLIEN MARDCGEYAH TRYFRGGMLT NARLLFGPTV RLPDLIIFLH TLNNIFEPHV AVRDAAKMNI PTVGIVDTNC NPCLITYPVP GNDDSPLAVH LYCRLFQTAI TRAKEKRQQV EALYRLQGQK EPGDQGPAHP PGADMSHSL | |
Greater than 85.0% as determined by SDS-PAGE. |