missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MRPS25 Protein
A cDNA sequence encoding the MRPS25 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12734-1mg
Additional Details : Weight : 0.01000kg
Specifications
MRPS25 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMPMKGRF PIRRTLQYLS QGNVVFKDSV KVMTVNYNTH GELGEGARKF VFFNIPQIQY KNPWVQIMMF KNMTPSPFLR FYLDSGEQVL VDVETKSNKE IMEHIRKILG KNEETLREEE EEKKQLSHPA NFGPRKYCLR ECICEVEGQV PCPSLVPLPK EMRGKYKAAL KADAQD | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The MRPS25 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |