missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MRRF Protein

A cDNA sequence encoding the MRRF was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12736-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15931379

  • 3915.00 € / 1mg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

MRRF Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MATKKAKAKG KGQSQTRVNI NAALVEDIIN LEEVNEEMKS VIEALKDNFN KTLNIRTSPG SLDKIAVVTA DGKLALNQIS QISMKSPQLI LVNMASFPEC TAAAIKAIRE SGMNLNPEVE GTLIRVPIPQ VTREHREMLV KLAKQNTNKA KDSLRKVRTN SMNKLKKSKD TVSEDTIRLI EKQISQMADD TVAELDRHLA VKTKELLG
Greater than 95.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
MRRF protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer, (pH 8.0), 0.2M NaCl, 30% glycerol and 2mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers