missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MTDH Protein
A cDNA sequence encoding the MTDH was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Name | MTDH Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15911419
|
enQuireBio™
QP12748-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15991409
|
enQuireBio™
QP12748-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15901419
|
enQuireBio™
QP12748-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
MTDH Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MTDH was filtered (0.4microm) and lyophilized in 20mM Tris buffer and 50mM NaCl, pH 7.5. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHASVSSGLNENLT VNGGGWNEKS VKLSSQISAG EEKWNSVSPA SAGKRKAEPS AWSQDTGDAN TNGKDWGRSW SDRSIFSGIG STAEPVSQST TSDYQWDVSR NQPYIDDEWS GLNGLSSADP NSDWNAPAEE WGNWVDEERA SLLKSQEPIP DDQKVSDDDK EKGEGALPTG KSKKKKKKKK KQGEDN | |
Greater than 90.0% as determined by SDS-PAGE. |