missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MTHFD2 Protein

A cDNA sequence encoding the MTHFD2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12749-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15931419

  • 203.00 € / 20µg
Estimated Shipment: 04-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

10797
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSLAAVRNE AVVISGRKLA QQIKQEVRQE VEEWVASGNK RPHLSVILVG ENPASHSYVL NKTRAAAVVG INSETIMKPA SISEEELLNL INKLNNDDNV DGLLVQLPLP EHIDERRICN AVSPDKDVDG FHVINVGRMC LDQYSMLPAT PWGVWEIIKR TGIPTLGKNV VVAGRSKNVG MPIAMLLHTD GAHERPGGDA TVTISHRYTP KEQLKKHTIL ADIVISAAGI PNLITADMIK EGAAVIDVGI NRVHDPVTAK PKLVGDVDFE GVRQKAGYIT PVPGGVGPMT VAMLMKNTII AAKKVLRLEE REVLKSKELG VATN
Greater than 95.0% as determined by SDS-PAGE.
MTHFD2 Protein
Research Use Only
MTHFD2
Human
His
The MTHFD2 solution (1 mg/ml) contains Phosphate buffered saline (pH 7.4), 20% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers