missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Mutant GSTO1 Protein
A cDNA sequence encoding the Mutant GSTO1 was constructed and used to recombinantly synthesize the protein.
131.00 € - 6525.00 €
Specifications
Name | Mutant GSTO1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15995429
|
enQuireBio™
QP14021-HIS-EC-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15975429
|
enQuireBio™
QP14021-HIS-EC-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15985429
|
enQuireBio™
QP14021-HIS-EC-1MG |
1 mg |
6525.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Mutant GSTO1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
GSTO1 protein is supplied in 1x PBS and 50% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MSGESARSLG KGSAPPGPVP EGSIRIYSMR FCPFAERTRL VLKAKGIRHE VININLKNKP EWFFKKNPFG LVPVLENSQG QLIYESAITC EYLDEAYPGK KLLPDDPYEK ACQKMILELF SKVPSLVGSF IRSQNKEDYA GLKEEFRKEF TKLEEVLTNK KTTFFGGNSI SMIDYLIWPW FERLEAMKLN ECVDHTPKLK LWMAAMKEDP TVSALLTSEK DWQGFLELYL QNSPEACDYG L | |
Greater than 95.0% as determined by SDS-PAGE. |