missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MYL6B Protein

A cDNA sequence encoding the MYL6B was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12772-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15961489

  • 3915.00 € / 1mg
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

MYL6B Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMPPKKDV PVKKPAGPSI SKPAAKPAAA GAPPAKTKAE PAVPQAPQKT QEPPVDLSKV VIEFNKDQLE EFKEAFELFD RVGDGKILYS QCGDVMRALG QNPTNAEVLK VLGNPKSDEL KSRRVDFETF LPMLQAVAKN RGQGTYEDYL EGFRVFDKEG NGKVMGAELR HVLTTLGEKM TEEEVETVLA GHEDSNGCIN YEAFLKHILS V
Greater than 95% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The MYL6B solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 2mM DTT and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers