missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MYL6B Protein
A cDNA sequence encoding the MYL6B was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12772-20ug
Additional Details : Weight : 0.01000kg
Specifications
MYL6B Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMPPKKDV PVKKPAGPSI SKPAAKPAAA GAPPAKTKAE PAVPQAPQKT QEPPVDLSKV VIEFNKDQLE EFKEAFELFD RVGDGKILYS QCGDVMRALG QNPTNAEVLK VLGNPKSDEL KSRRVDFETF LPMLQAVAKN RGQGTYEDYL EGFRVFDKEG NGKVMGAELR HVLTTLGEKM TEEEVETVLA GHEDSNGCIN YEAFLKHILS V | |
Greater than 95% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The MYL6B solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 2mM DTT and 10% glycerol. |