missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Myo Protein
A cDNA sequence encoding the Myo was constructed and used to recombinantly synthesize the protein.
131.00 € - 1305.00 €
Specifications
Gene ID (Entrez) | 4151 |
---|---|
Name | Myo Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | MB |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15971499
|
enQuireBio™
QP12775-20UG |
20 μg |
131.00 €
20µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15951499
|
enQuireBio™
QP12775-100UG |
100 μg |
203.00 €
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15961499
|
enQuireBio™
QP12775-1MG |
1 mg |
1305.00 €
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
4151 | |
Research Use Only | |
MB | |
Human | |
Untagged | |
The sterile solution contains phosphate-buffered saline (pH 8.0) and 50mM phosphate-borate. |
Myo Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGLSDGEWQL VLNVWGKVEA DIPGHGQEVL IRLFKGHPET LEKFDKFKHL KSEDEMKASE DLKKHGATVL TALGGILKKK GHHEAEIKPL AQSHATKHKI PVKYLEFISE CIIQVLQSKH PGDFGADAQG AMNKALELFR KDMASNYKEL GFQG | |
Greater than 95.0% as determined by SDS-PAGE. |