missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Myosin light chain 3 Protein

A DNA sequence encoding the Homo sapiens (Human) Myosin light chain 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6401-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16043050

  • 735.00 € / 100µg
Estimated Shipment: 04-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000249.1
Myosin light chain 3 Protein
Research Use Only
MYL3
Recombinant Protein
E. coli
APKKPEPKKD DAKAAPKAAP APAPPPEPER PKEVEFDASK IKIEFTPEQI EEFKEAFMLF DRTPKCEMKI TYGQCGDVLR ALGQNPTQAE VLRVLGKPRQ EELNTKMMDF ETFLPMLQHI SKNKDTGTYE DFVEGLRVFD KEGNGTVMGA ELRHVLATLG ERLTEDEVEK LMAGQEDSNG CINYEAFVKH IMSS
Greater than 80% as determined by SDS-PAGE.
4634
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers