missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Myosin regulatory light chain 2, atrial isoform Protein

A DNA sequence encoding the Homo sapiens (Human) Myosin regulatory light chain 2, atrial isoform, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8227-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16063401

  • 2285.00 € / 1mg
Estimated Shipment: 04-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_067046.1
Myosin regulatory light chain 2, atrial isoform Protein
Research Use Only
MYL7
Recombinant Protein
E. coli
MASRKAGTRG KVAATKQAQR GSSNVFSMFE QAQIQEFKEA FSCIDQNRDG IICKADLRET YSQLGKVSVP EEELDAMLQE GKGPINFTVF LTLFGEKLNG TDPEEAILSA FRMFDPSGKG VVNKDEFKQL LLTQADKFSP AEVEQMFALT PMDLAGNIDY KSLCYIITHG DEKEE
Greater than 80% as determined by SDS-PAGE.
58498
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers