missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human N6AMT1 / HEMK2 Protein
A DNA sequence encoding the Homo sapiens (Human) N6AMT1 / HEMK2, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_037372.4 |
---|---|
Gene ID (Entrez) | 29104 |
Name | N6AMT1 / HEMK2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16011220
|
enQuireBio™
QP6156-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16031220
|
enQuireBio™
QP6156-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16021220
|
enQuireBio™
QP6156-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16041220
|
enQuireBio™
QP6156-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16051220
|
enQuireBio™
QP6156-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16061220
|
enQuireBio™
QP6156-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_037372.4 | |
N6AMT1 / HEMK2 Protein | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
29104 | |
Research Use Only | |
N6AMT1 | |
Recombinant Protein | |
E. coli | |
MAGENFATPF HGHVGRGAFS DVYEPAEDTF LLLNALEAAA AELAGVEICL EVGSGSGVVS AFLASMIGPQ ALYMCTDINP EAAACTLETA RCNKVHIQPV ITDLVGSHGI EAAWAGGKNG REVMDRFFPL VPDLLSPKGL FYLVTIKENN PEEILKIMKT KGLQGTTALS RQAGQETLSV LKFTKS | |
Greater than 80% as determined by SDS-PAGE. |