missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NANOGP8 Protein
A cDNA sequence encoding the NANOGP8 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12788-2ug
Additional Details : Weight : 0.01000kg
Specifications
388112 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MSVDPACPQS LPCFEASDCK ESSPMPVICG PEENYPSLQM SSAEMPHTET VSPLPSSMDL LIQDSPDSST SPKGKQPTSA ENSVAKKEDK VPVKKQKTRT VFSSTQLCVL NDRFQRQKYL SLQQMQELSN ILNLSYKQVK TWFQNQRMKS KRWQKNNWPK NSNGVTQKAS APTYPSLYSS YHQGCLVNPT GNLPMWSNQT WNNSTWSNQT QNIQSWSNHS WNTQTWCTQS WNNQAWNSPF YNCGEESLQS CMHFQPNSPA SDLEAALEAA GEGLNVIQQT TRYFSTPQTM DLFLNYSMNM QPEDV | |
Greater than 85% as determined by SDS-PAGE. |
NANOGP8 Protein | |
Research Use Only | |
NANOGP8 | |
Human | |
Untagged | |
The NANOGP8 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 5% glycerol. |