missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NARS Protein

Product Code. 15901569
Change view
Click to view available options
Quantity:
10 μg
2 μg
Unit Size:
10µg
2µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15901569 2 μg 2µg
15991559 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15901569 Supplier enQuireBio™ Supplier No. QP12794HISINSECT2ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the NARS was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 4677
Name NARS Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol NARS
Product Type Recombinant Protein
Cross Reactivity Human
Species Baculovirus
Protein Tag His
Sequence GSSHHHHHH SSGLVPRGSH MGSMVLAELY VSDREGSDAT GDGTKEKPFK TGLKALMTVG KEPFPTIYVD SQKENERWNV ISKSQLKNIK KMWHREQMKS ESREKKEAED SLRREKNLEE AKKITIKNDP SLPEPKCVKI GALEGYRGQR VKVFGWVHRL RRQGKNLMFL VLRDGTGYLQ CVLADELCQC YNGVLLSTES SVAVYGMLNL TPKGKQAPGG HELSCDFWEL IGLAPAGGAD NLINEESDVD VQLNNRHMMI RGENMSKILK ARSMVTRCFR DHFFDRGYYE VTPPTLVQTQ VEGGATLFKL DYFGEEAFLT QSSQLYLETC LPALGDVFCI AQSYRAEQSR TRRHLAEYTH VEAECPFLTF DDLLNRLEDL VCDVVDRILK SPAGSIVHEL NPNFQPPKRP FKRMNYSDAI VWLKEHDVKK EDGTFYEFGE DIPEAPERLM TDTINEPILL CRFPVEIKSF YMQRCPEDSR LTESVDVLMP NVGEIVGGSM RIFDSEEILA GYKREGIDPT PYYWYTDQRK YGTCPHGGYG LGLERFLTWI LNRYHIRDVC LYPRFVQRCT P
Buffer NARS protein solution (0.25 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
Purity or Quality Grade Greater than 80.0% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.