missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NCR3 Protein
A cDNA sequence encoding the NCR3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12804-1mg
Additional Details : Weight : 0.01000kg
Specifications
NCR3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MLWVSQPPEI RTLEGSSAFL PCSFNASQGR LAIGSVTWFR DEVVPGKEVR NGTPEFRGRL APLASSRFLH DHQAELHIRD VRGHDASIYV CRVEVLGLGV GTGNGTRLVV EKEHPQLGAG T | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
NCR3 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |