missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NCR3 Protein

A cDNA sequence encoding the NCR3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12804-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15991589

  • 203.00 € / 20µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NCR3 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MLWVSQPPEI RTLEGSSAFL PCSFNASQGR LAIGSVTWFR DEVVPGKEVR NGTPEFRGRL APLASSRFLH DHQAELHIRD VRGHDASIYV CRVEVLGLGV GTGNGTRLVV EKEHPQLGAG T
Greater than 90% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
NCR3 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers