missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NDUFB5 Protein
A DNA sequence encoding the Homo sapiens (Human) NDUFB5, was expressed in the hosts and tags indicated.
160.00 € - 2285.00 €
Specifications
Accession Number | NP_001186886.1 |
---|---|
Gene ID (Entrez) | 4711 |
Name | NDUFB5 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16043140
|
enQuireBio™
QP6414-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16063140
|
enQuireBio™
QP6414-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16053140
|
enQuireBio™
QP6414-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16073140
|
enQuireBio™
QP6414-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16083140
|
enQuireBio™
QP6414-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16093140
|
enQuireBio™
QP6414-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Spécification
NP_001186886.1 | |
NDUFB5 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
4711 | |
Research Use Only | |
NDUFB5 | |
Recombinant Protein | |
E. coli | |
GQAELAEIPE GYVPEHWEYY KHPISRWIAR NFYDSPEKIY ERTMAVLQIE AEKAELRVKE LEVRKLMHVR GDGPWYYYET IDKELIDHSP KATPDN | |
Greater than 90% as determined by SDS-PAGE. |