missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Neuregulin-2/NRG2 Fc Chimera Protein
Description
Measured in a serum-free cell proliferation assay using MCF-7 human breast cancer cells. Karey, K.P. et al. (1988) Cancer Research 48:4083. The ED50 for this effect is 10.0-100 ng/mL.
Specifications
Specifications
| Accession Number | O14511.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 9542 |
| Molecular Weight (g/mol) | MolecularWeight-observed: 67-76 kDa, under reducing conditions., MolecularWeight-theroretical: 59 kDa |
| Quantity | 1 mg |
| Source | Human embryonic kidney cell, HEK293-derived human Neuregulin-2/NRG2 protein Human NRG2 (Cys112-Lys404) (N-terminus) Accession # O14511.1 GGIEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | divergent of neuregulin-1, Don-1, DON1, HRG2, neuregulin 2, Neuregulin2, NRG2, NTAK, NTAKneural- and thymus-derived activator for ErbB kinases, pro-neuregulin-2, membrane-bound isoform, Pro-NRG2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?