missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Neuregulin-2/NRG2 Fc Chimera Protein

Product Code. 18785825
Change view
Click to view available options
Quantity:
1 mg
100 μg
50 μg
Unit Size:
100µg
1mg
50µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18785825 100 μg 100µg
18766255 1 mg 1mg
18706005 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18785825 Supplier R&D Systems™ Supplier No. 11354NR100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Carrier Free Recombinant Protein

Measured in a serum-free cell proliferation assay using MCF-7 human breast cancer cells. Karey, K.P. et al. (1988) Cancer Research 48:4083. The ED50 for this effect is 10.0-100 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O14511.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 9542
Molecular Weight (g/mol) MolecularWeight-observed: 67-76 kDa, under reducing conditions., MolecularWeight-theroretical: 59 kDa
Quantity 100 μg
Source Human embryonic kidney cell, HEK293-derived human Neuregulin-2/NRG2 protein Human NRG2 (Cys112-Lys404) (N-terminus) Accession # O14511.1 GGIEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias divergent of neuregulin-1, Don-1, DON1, HRG2, neuregulin 2, Neuregulin2, NRG2, NTAK, NTAKneural- and thymus-derived activator for ErbB kinases, pro-neuregulin-2, membrane-bound isoform, Pro-NRG2
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Neuregulin-2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.