missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NEURL2 Protein
A cDNA sequence encoding the NEURL2 was constructed and used to recombinantly synthesize the protein.
131.00 € - 7540.00 €
Specifications
Name | NEURL2 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15931689
|
enQuireBio™
QP12831-2UG |
2 μg |
131.00 €
2µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15911689
|
enQuireBio™
QP12831-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15921689
|
enQuireBio™
QP12831-1MG |
1 mg |
7540.00 €
1mg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NEURL2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The NEURL2 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAAASEP VDSGALWGLE RPEPPPTRFH RVHGANIRVD PSGTRATRVE SFAHGVCFSR EPLAPGQVFL VEIEEKELGW CGHLRLGLTA LDPASLAPVP EFSLPDLVNL GHTWVFAITR HHNRVPREGR PEAEAAAPSR PPTLLVEPYL RIEQFRIPRDRLVGRSRPGL YSHLLDQLYE LNVLPPTARR SRLGVLFCPR PDGTADMHII INGEDMGPSA RGLPAAQPLY AVVDVFASTK SVRLVQLEYG LPSLQTLCRL VIQRSMVHRL AIDGLHLPKE LKDFCKYE | |
Greater than 85.0% as determined by SDS-PAGE. |