missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Neutrophil Elastase/ELA2 Protein

Product Code. 18799376
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18799376 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18799376 Supplier R&D Systems™ Supplier No. 9167SE020
  • ONLINE EXCLUSIVE PRICE
    644.56 €
    571.20 € / 20µg
    Save 73.36 €
    11% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

The Recombinant Human Neutrophil Elastase/ELA2 Protein is derived from CHO. The Recombinant Human Neutrophil Elastase/ELA2 Protein has been validated for the following applications: Enzyme Activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P08246
For Use With (Application) Enzyme Activity
Gene ID (Entrez) 1991
Molecular Weight (g/mol) Observed M.W.: 29-39kDa, reducing conditions, Theoretical M.W.: 25kDa
Quantity 20 μg
Source Chinese Hamster Ovary cell line,CHO-derived human Neutrophil Elastase/ELA2 protein Ser28-Asn252,with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU per 1μg of the protein by the LAL method.
Gene Alias Bone marrow serine protease, EC 3.4.21, EC 3.4.21.37, ELA2, ELA2granulocyte-derived elastase, ELANE, elastase 2, neutrophil, elastase, neutrophil expressed, Elastase-2, GE, HLEelastase-2, HNE, Human leukocyte elastase, Leukocyte Elastase, medullasin, NE, neutrophil elastase, PMN elastase, PMN-E, polymorphonuclear elastase, SCN1
Common Name Neutrophil Elastase/ELA2
Biological Activity Measured by its ability to cleave the fluorogenic peptide substrate, MeOSuc-Ala-Ala-Pro-Val-7-amido-4-methylcoumarin (MeOSuc-AAPV-AMC). The specific activity is >1,500 pmol/min/μg, as measured under the described conditions. See Activity Assay Protocol on RnDSystems.com.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Ser28-Asn252, with a C-terminal 10-His tag
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.