missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NGFR Protein
A cDNA sequence encoding the NGFR was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12841-1mg
Specifications
NGFR Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
KEACPTGLYT HSGECCKACN LGEGVAQPCG ANQTVCEPCL DSVTFSDVVS ATEPCKPCTE CVGLQSMSAP CVEADDAVCR CAYGYYQDET TGRCEACRVC EAGSGLVFSC QDKQNTVCEE CPDGTYSDEA NHVDPCLPCT VCEDTERQLR ECTRWADAEC EEIPGRWITR STPPEGSDST APSTQEPEAP PEQDLIASTV AGVVTTVMGS SQPVVTRGTT DNLEHHHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
NGFR protein solution (1 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |