missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NMI Protein
A cDNA sequence encoding the NMI was constructed and used to recombinantly synthesize the protein.
131.00 € - 3915.00 €
Specifications
Name | NMI Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15911769
|
enQuireBio™
QP12857-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15901769
|
enQuireBio™
QP12857-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15991759
|
enQuireBio™
QP12857-1MG |
1 mg |
3915.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NMI Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The NMI protein solution (0.5 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0)1mM DTT and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MEADKDDTQQ ILKEHSPDEF IKDEQNKGLI DEITKKNIQLK KEIQKLETE LQEATKEFQI KEDIPETKMK FLSVETPEND SQLSNISCSF QVSSKVPYEI QKGQALITFE KEEVAQNVVS MSKHHVQIKDV NLEVTAKPV PLNSGVRFQV YVEVSKMKIN VTEIPDTLRE DQMRDKLELS FSKSRNGGGE VDRVDYDRQS GSAVITFVEI GVADKILKKKEYPLYINQTC HRVTVSPYTE IHLKKYQIFS GTSKRTVLLT GMEGIQMDEE IVEDLINIHF QRAKNGGGEV DVVKCSLGQP HIAYFEE | |
Greater than 90% as determined by SDS-PAGE. |