missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NQO1 Protein
A cDNA sequence encoding the NQO1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12875-10ug
Additional Details : Weight : 0.01000kg
Specifications
1728 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MVGRRALIVL AHSERTSFNY AMKEAAAAAL KKKGWEVVES DLYAMNFNPI ISRKDITGKL KDPANFQYPA ESVLAYKEGH LSPDIVAEQK KLEAADLVIF QFPLQWFGVP AILKGWFERV FIGEFAYTYA AMYDKGPFRS KKAVLSITTG GSGSMYSLQG IHGDMNVILW PIQSGILHFC GFQVLEPQLT YSIGHTPADA RIQILEGWKK RLENIWDETP LYFAPSSLFD LNFQAGFLMK KEVQDEEKNK KFGLSVGHHL GKSIPTDNQI KARK | |
Greater than 95.0% as determined by SDS-PAGE. |
NQO1 Protein | |
Research Use Only | |
NQO1 | |
Human | |
His | |
The NQO1 solution contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 1mM DTT. |