missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NR1I3 Protein
A DNA sequence encoding the Homo sapiens (Human) NR1I3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7599-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001070937.1 | |
NR1I3 Protein | |
Research Use Only | |
NR1I3 | |
Recombinant Protein | |
E. coli | |
ASREDELRNC VVCGDQATGY HFNALTCEGC KGFFRRTVSK SIGPTCPFAG SCEVSKTQRR HCPACRLQKC LDAGMRKDMI LSAEALALRR AKQAQRRAQQ TPVQLSKEQE ELIRTLLGAH TRHMGTMFEQ FVQFRPPAHL FIHHQPLPTL APVLPLVTHF ADINTFMVLQ VIKFTKDLPV FRSLPIEDQI SLLKGAAVEI CHIVLNTTFC LQTQNFLCGP LRYTIEDGAR VSPTVGFQVE FLELLFHFHG TLRKLQLQEP EYVLLAAMAL FSPDRPGVTQ RDEIDQLQEE MALTLQSYIK GQQRRPRDRF LYAKLLGLLA ELRSINEAYG YQIQHIQGLS AMMPLLQEIC | |
Greater than 90% as determined by SDS-PAGE. |
9970 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |