missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NRG4 Protein
A cDNA sequence encoding the NRG4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12880-2ug
Additional Details : Weight : 0.01000kg
Specifications
NRG4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMPTDHEE PCGPSHKSFC LNGGLCYVIP TIPSPFCRCV ENYTGARCEE VFLPGSSIQT KSNL | |
Greater than 90% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
NRG4 protein solution (0.25 mg/ml) contains 20mM Tris-HCl(pH 8.0), 30% glycerol, 0.15M NaCl and 1mM DTT. |