missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NRGN Protein
A cDNA sequence encoding the NRGN was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12881-2ug
Additional Details : Weight : 0.01000kg
Specifications
NRGN Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMDCCTEN ACSKPDDDIL DIPLDDPGAN AAAAKIQASF RGHMARKKIK SGERGRKGPG PGGPGGAGVA RGGAGGGPSG D | |
Greater than 90% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
NRGN protein solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 7.0), 30% glycerol 0.1mM PMSF and 1mM EDTA. |