missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NT5C3B Protein
A cDNA sequence encoding the NT5C3B was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12889-20ug
Additional Details : Weight : 0.01000kg
Specifications
NT5C3B Protein | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
NT5C3B protein solution (1 mg/ml) containing Phosphate Buffered Saline (pH 7.4), 20% glycerol and 1mM DTT. |
Greater than 85.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAEEVST LMKATVLMRQ PGRVQEIVGA LRKGGGDRLQ VISDFDMTLS RFAYNGKRCP SSYNILDNSK IISEECRKEL TALLHHYYPI EIDPHRTVKE KLPHMVEWWT KAHNLLCQQK IQKFQIAQVV RESNAMLREG YKTFFNTLYH NNIPLFIFSA GIGDILEEII RQMKVFHPNI HIVSNYMDFN EDGFLQGFKG QLIHTYNKNS SACENSGYFQ QLEGKTNVIL LGDSIGDLTM ADGVPGVQNI LKIGFLNDKV EERRERYMDS YDIVLEKDET LDVVNGLLQH ILCQGVQLEM QGP |