missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Nucleolar transcription factor 1 Protein

Product Code. 16059151
Click to view available options
Quantity:
10 μg
100 μg
50 μg
Unit Size:
100µg
10µg
50µg
This item is not returnable. View return policy

Product Code. 16059151

Brand: enQuireBio™ QP9990iv100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens(Human) Nucleolar transcription factor 1, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001070151.1
Gene ID (Entrez) 7343
Name Nucleolar transcription factor 1 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol UBTF
Protein Length Full length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MNGEADCPTD LEMAAPKGQD RWSQEDMLTL LECMKNNLPS NDSSKFKTTE SHMDWEKVAF KDFSGDMCKL KWVEISNEVR KFRTLTELIL DAQEHVKNPY KGKKLKKHPD FPKKPLTPYF RFFMEKRAKY AKLHPEMSNL DLTKILSKKY KELPEKKKMK YIQDFQREKQ EFERNLARFR EDHPDLIQNA KKSDIPEKPK TPQQLWYTHE KKVYLKVRPD ATTKEVKDSL GKQWSQLSDK KRLKWIHKAL EQRKEYEEIM RDYIQKHPEL NISEEGITKS TLTKAERQLK DKFDGRPTKP PPNSYSLYCA ELMANMKDVP STERMVLCSQ QWKLLSQKEK DAYHKKCDQK KKDYEVELLR FLESLPEEEQ QRVLGEEKML NINKKQATSP ASKKPAQEGG KGGSEKPKRP VSAMFIFSEE KRRQLQEERP ELSESELTRL LARMWNDLSE KKKAKYKARE AALKAQSERK PGGEREERGK LPESPKRAEE IWQQSVIGDY LARFKNDRVK ALKAMEMTWN NMEKKEKLMW IKKAAEDQKR YERELSEMRA PPAATNSSKK MKFQGEPKKP PMNGYQKFSQ ELLSNGELNH LPLKERMVEI GSRWQRISQS QKEHYKKLAE EQQKQYKVHL DLWVKSLSPQ DRAAYKEYIS NKRKSMTKLR GPNPKSSRTT LQSKSESEED DEEDEDDEDE DEEEEDDENG DSSEDGGDSS ESSSEDESED GDENEEDDED EDDDEDDDED EDNESEGSSS SSSSSGDSSD SDSN
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 85% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.