missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NUDT16 Protein

A cDNA sequence encoding the NUDT16 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12902-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15951899

  • 203.00 € / 20µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NUDT16 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MAGARRLELG EALALGSGWR HACHALLYAP DPGMLFGRIP LRYAILMQMR FDGRLGFPGG FVDTQDRSLE DGLNRELREE LGEAAAAFRV ERTDYRSSHV GSGPRVVAHF YAKRLTLEEL LAVEAGATRA KDHGLEVLGL VRVPLYTLRD GVGGLPTFLE NSFIGSAREQ LLEALQDLGL LQSGSISGLK IPAHH
Greater than 95% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The NUDT16 protein solution (1 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0) 1mM DTT, 100mM NaCl and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers