missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NUDT4 Protein
A cDNA sequence encoding the NUDT4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12906-10ug
Additional Details : Weight : 0.01000kg
Specifications
NUDT4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMMKFKPN QTRTYDREGF KKRAACLCFR SEQEDEVLLV SSSRYPDQWI VPGGGMEPEE EPGGAAVREV YEEAGVKGKL GRLLGIFENQ DRKHRTYVYV LTVTEILEDW EDSVNIGRKR EWFKVEDAIK VLQCHKPVHA EYLEKLKLGC SPANGNSTVP SLPDNNALFV TAAQTSGLPS SVR | |
Greater than 90.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
NUDT4 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.1M NaCl. |