missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human O-GlcNAcTransferase/OGT Protein

Product Code. 18736417
Change view
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18736417 10 μg 10µg
18747407 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18736417 Supplier R&D Systems™ Supplier No. 8446GT010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to transfer GlcNAc from UDP-GclNAc to peptide OGT substrate from AnaSpec, Inc. The specific activity is >25 pmol/min/μg, as measured under the described conditions.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O15294
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris, NaCl and TCEP.
Gene ID (Entrez) 8473
Molecular Weight (g/mol) M.W. (Observed): 70 kDa, reducing conditions; M.W. (theoretical): 81 kDa
Name O-GlcNAc Transferase/OGT
Quality Control Testing SDS-PAGE: 70kDa, reducing conditions
Quantity 10 μg
Source E. coli-derived human O-GlcNAc Transferase/OGT protein Cys323-Glu1041, with C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Regulatory Status RUO
Termination Cys323
Without Additives Carrier Protein
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Alias EC 2.4.1, EC 2.4.1.255 FLJ23071, HRNT1, MGC22921, O-GLCNAC, OGlcNAc Transferase, O-GlcNAc transferase p110 subunit, O-GlcNAc transferase subunit p110, OGT, O-linked N-acetylglucosamine (GlcNAc) transferase(UDP-N-acetylglucosamine:polypeptide-N-acetylglucosaminyl transferase), O-linked N-acetylglucosamine transferase 110 kDa subunit, UDP-N-acetylglucosamine--peptide N-acetylglucosaminyltransferase 110 kDasubunit, uridinediphospho-N-acetylglucosamine:polypeptide beta-N-acetylglucosaminyltransferase
Gene Symbol OGT
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >80%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein O-GlcNAc Transferase p110 subunit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.