missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human O-GlcNAcTransferase/OGT Protein
Description
Measured by its ability to transfer GlcNAc from UDP-GclNAc to peptide OGT substrate from AnaSpec, Inc. The specific activity is >25 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | O15294 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl and TCEP. |
| Gene ID (Entrez) | 8473 |
| Molecular Weight (g/mol) | M.W. (Observed): 70 kDa, reducing conditions; M.W. (theoretical): 81 kDa |
| Name | O-GlcNAc Transferase/OGT |
| Quality Control Testing | SDS-PAGE: 70kDa, reducing conditions |
| Quantity | 10 μg |
| Source | E. coli-derived human O-GlcNAc Transferase/OGT protein Cys323-Glu1041, with C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?