missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human OBFC1 Protein

A cDNA sequence encoding the OBFC1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12917-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15921949

  • 203.00 € / 10µg
Estimated Shipment: 04-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

OBFC1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMQPGSSR CEEETPSLLW GLDPVFLAFA KLYIRDILDM KESRQVPGVF LYNGHPIKQV DVLGTVIGVR ERDAFYSYGV DDSTGVINCI CWKKLNTESV SAAPSAAREL SLTSQLKKLQ ETIEQKTKIE IGDTIRVRGS IRTYREEREI HATAYYKVDD PVWNIQIARM LELPTIYRKV YDQPFHSSAL EKEEALSNPG ALDLPSLTSL LSEKAKEFLM ENRVQSFYQQ ELEMVESLLS LANQPVIHSA CSDQVNFKKD TTSKAIHSIF KNAIQLLQEK GLVFQKDDGF DNLYYVTRED KDLHRKIHRI IQQDCQKPNH MEKGCHFLHI LACARLSIRP GLSEAVLQQV LELLEDQSDI VSTMEHYYTA F
Greater than 85% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The OBFC1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers