missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human OBFC1 Protein
A cDNA sequence encoding the OBFC1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12917-1mg
Specifications
OBFC1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMQPGSSR CEEETPSLLW GLDPVFLAFA KLYIRDILDM KESRQVPGVF LYNGHPIKQV DVLGTVIGVR ERDAFYSYGV DDSTGVINCI CWKKLNTESV SAAPSAAREL SLTSQLKKLQ ETIEQKTKIE IGDTIRVRGS IRTYREEREI HATAYYKVDD PVWNIQIARM LELPTIYRKV YDQPFHSSAL EKEEALSNPG ALDLPSLTSL LSEKAKEFLM ENRVQSFYQQ ELEMVESLLS LANQPVIHSA CSDQVNFKKD TTSKAIHSIF KNAIQLLQEK GLVFQKDDGF DNLYYVTRED KDLHRKIHRI IQQDCQKPNH MEKGCHFLHI LACARLSIRP GLSEAVLQQV LELLEDQSDI VSTMEHYYTA F | |
Greater than 85% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The OBFC1 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |