missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human OLFM1 Protein

Product Code. p-7150803
Change view
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15921969 2 μg 2µg
15901969 10 μg 10µg
15911969 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15921969 Supplier enQuireBio™ Supplier No. QP129232ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the OLFM1 was constructed and used to recombinantly synthesize the protein.

Specifications

Name OLFM1 Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MGSLPTNPEE SWQVYSSAQD SEGRCICTVV APQQTMCSRD ARTKQLRQLL EKVQNMSQSI EVLDRRTQRD LQYVEKMENQ MKGLESKFKQ VEESHKQHLA RQFKG
Buffer OLFM1 protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer, (pH 7.5), 0.2M NaCl, 50% glycerol and 5mM DTT.
Purity or Quality Grade Greater than 90.0% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.