missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human OSGEP Protein

A DNA sequence encoding the Homo sapiens (Human) OSGEP, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8150-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16032921

  • 2285.00 € / 1mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_060277.1
OSGEP Protein
Research Use Only
OSGEP
Recombinant Protein
E. coli
MPAVLGFEGS ANKIGVGVVR DGKVLANPRR TYVTPPGTGF LPGDTARHHR AVILDLLQEA LTESGLTSQD IDCIAYTKGP GMGAPLVSVA VVARTVAQLW NKPLVGVNHC IGHIEMGRLI TGATSPTVLY VSGGNTQVIA YSEHRYRIFG ETIDIAVGNC LDRFARVLKI SNDPSPGYNI EQMAKRGKKL VELPYTVKGM DVSFSGILSF IEDVAHRMLA TGECTPEDLC FSLQETVFAM LVEITERAMA HCGSQEALIV GGVGCNVRLQ EMMATMCQER GARLFATDER FCIDNGAMIA QAGWEMFRAG HRTPLSDSGV TQRYRTDEVE VTWRD
Greater than 80% as determined by SDS-PAGE.
55644
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers