missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human OXLD1 Protein
A cDNA sequence encoding the OXLD1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12941-10ug
Additional Details : Weight : 0.01000kg
Spezifikation
339229 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSPGAQAPD GRRKFGTDHV EVGSQAGADG TRPPKASLPP ELQPPTNCCM SGCPNCVWVE YADRLLQHFQ DGGERALAAL EEHVADENLK AFLRMEIRLH TRCGG | |
Greater than 90.0% as determined by SDS-PAGE. |
OXLD1 Protein | |
Research Use Only | |
OXLD1 | |
Human | |
His | |
OXLD1 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 50% glycerol and 1mM DTT. |