missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human p38 delta / MAPK13 Protein

A DNA sequence encoding the Homo sapiens (Human) p38 delta / MAPK13, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6339-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16042590

  • 2285.00 € / 1mg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_002745.1
p38 delta / MAPK13 Protein
Research Use Only
MAPK13
Recombinant Protein
E. coli
MSLIRKKGFY KQDVNKTAWE LPKTYVSPTH VGSGAYGSVC SAIDKRSGEK VAIKKLSRPF QSEIFAKRAY RELLLLKHMQ HENVIGLLDV FTPASSLRNF YDFYLVMPFM QTDLQKIMGM EFSEEKIQYL VYQMLKGLKY IHSAGVVHRD LKPGNLAVNE DCELKILDFG LARHADAEMT GYVVTRWYRA PEVILSWMHY NQTVDIWSVG CIMAEMLTGK TLFKGKDYLD QLTQILKVTG VPGTEFVQKL NDKAAKSYIQ SLPQTPRKDF TQLFPRASPQ AADLLEKMLE LDVDKRLTAA QALTHPFFEP FRDPEEETEA QQPFDDSLEH EKLTVDEWKQ HIYKEIVNFS PIARKDSRRR SGMKL
Greater than 90% as determined by SDS-PAGE.
5603
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers