missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human P4HB / ERBA2L Protein

Product Code. 16073450
Click to view available options
Quantity:
1 mg
10 μg
200 μg
50 μg
500 μg
Unit Size:
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16073450

Brand: enQuireBio™ QP6460ec1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) P4HB / ERBA2L, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_000909.2
Gene ID (Entrez) 5034
Name P4HB / ERBA2L Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol P4hb
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence DAPEEEDHVL VLRKSNFAEA LAAHKYLLVE FYAPWCGHCK ALAPEYAKAA GKLKAEGSEI RLAKVDATEE SDLAQQYGVR GYPTIKFFRN GDTASPKEYT AGREADDIVN WLKKRTGPAA TTLPDGAAAE SLVESSEVAV IGFFKDVESD SAKQFLQAAE AIDDIPFGIT SNSDVFSKYQ LDKDGVVLFK KFDEGRNNFE GEVTKENLLD FIKHNQLPLV IEFTEQTAPK IFGGEIKTHI LLFLPKSVSD YDGKLSNFKT AAESFKGKIL FIFIDSDHTD NQRILEFFGL KKEECPAVRL ITLEEEMTKY KPESEELTAE RITEFCHRFL EGKIKPHLMS QELPEDWDKQ PVKVLVGKNF EDVAFDEKKN VFVEFYAPWC GHCKQLAPIW DKLGETYKDH ENIVIAKMDS TANEVEAVKV HSFPTLKFFP ASADRTVIDY NGERTLDGFK KFLESGGQDG AGDDDDLEDL EEAEEPDMEE DDDQKAVKDE L
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.