missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PA28A / PSME1 Protein

A DNA sequence encoding the Homo sapiens (Human) PA28A / PSME1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6547-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16034070

  • 518.00 € / 100µg
Estimated Shipment: 04-08-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001268457.1
PA28A / PSME1 Protein
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
5720
Greater than 80% as determined by SDS-PAGE.
Research Use Only
PSME1
Recombinant Protein
E. coli
MAMLRVQPEA QAKVDVFRED LCTKTENLLG SYFPKKISEL DAFLKEPALN EANLSNLKAP LDIPVPDPVK EKEKEERKKQ QEKEDKDEKK KGEDEDKGPP CGPVNCNEKI VVLLQRLKPE IKDVIEQLNL VTTWLQLQIP RIEDGNNFGV AVQEKVFELM TSLHTKLEGF HTQISKYFSE RGDAVTKAAK QPHVGDYRQL VHELDEAEYR DIRLMVMEIR NAYAVLYDII LKNFEKLKKP RGETKGMIY
Videos
SDS
Documents
Certificates
Special Offers

Special Offers