missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PAX9 Protein
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
Specifications
Specifications
| Name | PAX9 Protein |
| Quantity | 2 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSMEPAFGE VNQLGGVFVN GRPLPNAIRL RIVELAQLGI RPCDISRQLR VSHGCVSKIL ARYNETGSIL PGAIGGSKPR VTTPTVVKHI RTYKQRDPGI FAWEIRDRLL ADGVCDKYNV PSVSSISRIL RNKIGNLAQQ GHYDSYKQHQ PTPQPALPYN HIYSYPSPIT AAAAKVPTPP GVPAIPGSVA MPRTWPSSHS VTDILGIRSI TDQVSDSSPY HSPKVEEWSS LGRNNFPAAA PHAVNGLEKG ALEQEAKYGQ APNGLPAVGS FVSASSMAPY PTPAQVSPYM TYSAAPSGYV AGHGWQHAGG TSLSPHNCDI PASLAFKGMQ AAREGSHSVT ASAL |
| Buffer | PAX9 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction