missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PBEF / Visfatin / NAMPT Protein
Recombinant Protein
Brand: enQuireBio™ QP5362-5ug
Additional Details : Weight : 0.01000kg
Specifications
NP_005737.1 | |
PBEF / Visfatin / NAMPT Protein | |
5 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Human PBEF / Visfatin / NAMPT induces IL-6 production in human PBMCs as detected via ELISA (typical ED50 is 100 ng/ml). | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
10135 | |
Greater than 90 as determined by SDS-PAGE. | |
Research Use Only | |
NAMPT | |
Recombinant Protein | |
E. coli | |
MPPNTSKVYS YFECREKKTE NSKLRKVKYE ETVFYGLQYI LNKYLKGKVV TKEKIQEAKD VYKEHFQDDV FNEKGWNYIL EKYDGHLPIE IKAVPEGFVI PRGNVLFTVE NTDPECYWLT NWIETILVQS WYPITVATNS REQKKILAKY LLETSGNLDG LEYKLHDFGY RGVSSQETAG IGASAHLVNF KGTDTVAGLA LIKKYYGTKD PVPGYSVPAA EHSTITAWGK DHEKDAFEHI VTQFSSVPVS VVSDSYDIYN ACEKIWGEDL RHLIVSRSTQ APLIIRPDSG NPLDTVLKVL EILGKKFPVT ENSKGYKLLP PYLRVIQGDG VDINTLQEIV EGMKQKMWSI ENIAFGSGGG LLQKLTRDLL NCSFKCSYVV TNGLGINVFK DPVADPNKRS KKGRLSLHRT PAGNFVTLEE GKGDLEEYGQ DLLHTVFKNG KVTKSYSFDE IRKNAQLNIE LEAAHH |