Learn More
enQuireBio™ Recombinant Human PBEF / Visfatin / NAMPT Protein
Recombinant Protein
Brand:  enQuireBio™ QP5362-25ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15935979
  • 252.00 € / 25µg
Estimated Shipment: 21-06-2024
to see stock.
Specifications
NP_005737.1
PBEF / Visfatin / NAMPT Protein
Research Use Only
NAMPT
Recombinant Protein
E. coli
MPPNTSKVYS YFECREKKTE NSKLRKVKYE ETVFYGLQYI LNKYLKGKVV TKEKIQEAKD VYKEHFQDDV FNEKGWNYIL EKYDGHLPIE IKAVPEGFVI PRGNVLFTVE NTDPECYWLT NWIETILVQS WYPITVATNS REQKKILAKY LLETSGNLDG LEYKLHDFGY RGVSSQETAG IGASAHLVNF KGTDTVAGLA LIKKYYGTKD PVPGYSVPAA EHSTITAWGK DHEKDAFEHI VTQFSSVPVS VVSDSYDIYN ACEKIWGEDL RHLIVSRSTQ APLIIRPDSG NPLDTVLKVL EILGKKFPVT ENSKGYKLLP PYLRVIQGDG VDINTLQEIV EGMKQKMWSI ENIAFGSGGG LLQKLTRDLL NCSFKCSYVV TNGLGINVFK DPVADPNKRS KKGRLSLHRT PAGNFVTLEE GKGDLEEYGQ DLLHTVFKNG KVTKSYSFDE IRKNAQLNIE LEAAHH
Greater than 90 as determined by SDS-PAGE.
10135
25 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Human PBEF / Visfatin / NAMPT induces IL-6 production in human PBMCs as detected via ELISA (typical ED50 is 100 ng/ml).
Human
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA